Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Celatom∣r FW-14

Catalog No. AAH317330B
Click to view available options
Quantity:
250 g
1000 g

CAS: 68855-54-9 | CNa2O3 | 105.988 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
MDL Number MFCD00132803
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]

Specifications

Quantity 1000 g
Chemical Name or Material Celatom™ FW-14
Hazard Category Warning
Hazard Statement GHS H Statement
H351-H373
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
P261-P280h
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
EINECSNumber 272-489-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.