Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Celecoxib, 98%, Thermo Scientific Chemicals

Catalog No. AC456451000
Click to view available options
Quantity:
100 mg

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 169590-42-5
Molecular Formula C17H14F3N3O2S
Molecular Weight (g/mol) 381.37
InChI Key RZEKVGVHFLEQIL-UHFFFAOYSA-N
IUPAC Name 4-[5-(4-methylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzene-1-sulfonamide
SMILES CC1=CC=C(C=C1)C1=CC(=NN1C1=CC=C(C=C1)S(N)(=O)=O)C(F)(F)F

Specifications

Melting Point 156.0°C to 157.0°C
Assay Percent Range 98%
Quantity 100 mg
Formula Weight 381.37
Chemical Name or Material Celecoxib
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May damage the unborn child.

Precautionary Statement GHS P Statement Obtain special instructions before use.

Use personal protective equipment as required.

IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.