Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CELF3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989769
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9769 Supplier Bioss Supplier No. BS13830R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNA-binding protein involved in the regulation of pre- mRNA alternative splicing. Mediates exon inclusion and/or exclusion in pre-mRNA that are subject to tissue-specific and developmentally regulated alternative splicing. Specifically activates exon 5 inclusion of cardiac isoforms of TNNT2 during heart remodeling at the juvenile to adult transition. Activates the splicing of MAPT/Tau exon 10. Binds to muscle-specific splicing enhancer (MSE) intronic sites flanking the alternative exon 5 of TNNT2 pre-mRNA.

Specifications

Antigen CELF3
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene CELF3
Gene Accession No. Q5SZQ8
Gene Alias 4930415M08Rik; BRUNOL1; Bruno-like protein 1; CAG repeat domain; CAG repeat protein 4; CAGH4; Celf3; CELF-3; CUG-BP and ETR-3 like factor 3; CUG-BP- and ETR-3-like factor 3; CUGBP Elav-like family member 3; CUGBP, Elav-like family member 3; ELAV-type RNA-binding protein 1; ERDA4; ETR-1; Expanded repeat domain protein CAG/CTG 4; expanded repeat domain, CAG/CTG 4; RNA-binding protein BRUNOL-1; Tnrc4; trinucleotide repeat containing 4; Trinucleotide repeat-containing gene 4 protein; VCUGBP, Elav-like family member 3
Gene Symbols CELF3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CELF3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11189
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.