Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Celite™ 545, Thermo Scientific Chemicals

Catalog Number AC349670025
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Fiber Drum
Plastic Bottle
Plastic Drum
68855-54-9
CNa2O3
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
Calcinated diatomaceous earth, Diatomite
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]
68855-54-9
CNa2O3
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
Calcinated diatomaceous earth, Diatomite
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 2.5 kg
Density 336 g/L (typically)
Color White
pH 10.0 (10% aq. soln., typically)
Loss on Ignition 0.2% (600°C) (typically)
Particle Size 38.9 Micron
Packaging Plastic Drum
Permeability 4.20 Darcies (typically)
Silicon Dioxide 91.5%
Formula Weight 60.08
Chemical Name or Material Celite™ 545
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit