Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Celite™ 545, ACROS Organics™

Click to view available options
Quantity:
500g
2.5kg
10kg
Packaging:
Fibre drum
Plastic bottle
Plastic drum
68855-54-9
CNa2O3
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
Calcinated diatomaceous earth,Diatomite
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]
68855-54-9
CNa2O3
105.988
CDBYLPFSWZWCQE-UHFFFAOYSA-L
Calcinated diatomaceous earth,Diatomite
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]
Filter aid
Color White
pH 10.0 (10% aq. soln., typically)
Quantity 500g
Aluminum Oxide 0.01
Calcium Oxide 0.3%
Fe2O3 1.50%
Loss on Ignition 0.2% (600°C) (typically)
Magnesium Oxide 0.30%
Particle Size 38.9 Micron
Packaging Plastic bottle
Permeability 4.20 Darcies (typically)
Silicon Dioxide 91.5%
Sodium Oxide and Potassium Oxide Na2O + K2O: 2.5%
Wet Density 336g/L (typically)
Formula Weight 60.08
Physical Form Powder
Chemical Name or Material Celite™ 545
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning