Learn More
R&D Systems™ Cell Activation Cocktail 500X
GSA_VAGREENER_CHOICESDP
Catalog No. 54761ML Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
54-761-ML 1 mL
1 options
This item is not returnable. View return policy
Catalog No. 54-761-ML Supplier R&D Systems™ Supplier No. 5476/1ML
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. Cell activation cocktail
Cell Activation Cocktail 500X contains the following compounds: Monensin sodium salt (1.5 mM), Phorbol 12-myristate 13-acetate (0.0405 mM) and Ionomycin calcium salt (0.67 mM). Cocktail is provided as 1 ml of 500X concentrate in ethanol.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 1 mL
Product Line Tocris
Target Buffers, Chelators and Reagents