Learn More
Tocris Bioscience™ Cell counting kit-8
GREENER_CHOICESDP
Catalog No. 73685ML Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
73-685-ML 5 mL
1 options
This item is not returnable. View return policy
Catalog No. 73-685-ML Supplier Tocris Bioscience™ Supplier No. 7368/5ML
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cell viability and proliferation assay test solution
Cell Counting Kit-8 is a viability and proliferation assay test solution. It is a supplied as a ready-to-use 1-bottle solution and enables the colorimetric determination of the number of viable cells in cell proliferation and cytotoxicity assays. Cell Counting Kit-8 utilizes a highly water-soluble tetrazolium salt (WST-8), which is reduced by cellular dehydrogenases to an orange formazan dye. The color change can be detected in the 430 - 490 nm range and Cell Counting Kit-8 exhibits improved sensitivity compared with other tetrazolium salt-based assays such as MTT, XTT or MTS. Can also be used in the detection of superoxide.
For more information on how to use Cell Counting Kit-8, please see our protocol .
Specifications
Quantity 5 mL
Target Cell Viability/Proliferation Assays