Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Cell Culture 1-Tray, 630 cm⊃2

Catalog No. MP096143001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. per Pack:
1
14
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
MP096143001 14
MP096143011 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP096143001 Supplier MP Biomedicals™ Supplier No. 096143001
Only null left
Add to Cart
Add to Cart

Meticulously engineered cell culture trays suitable for efficient and consistent commercial scale-up for the bioproduction of monoclonal antibodies, extracellular vesicles, vaccines, stem cells, and enzymes.

  • Graduated on both sides
  • Tissue culture treated surface allows for effective cell adhesion and growth
  • Comes with closed and filtered caps
  • Leak-free
  • Irradiation sterilized
  • Pyrogen-free
  • DNase-free, RNase-free
TRUSTED_SUSTAINABILITY

Specifications

No. per Pack 14
Sterility Sterile
Culture Area 630 cm²

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.