Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Cell Culture, 10-Tray, 6310 cm⊃2
Shop All MP Biomedicals, Inc Products
Click to view available options
No. per Case:
20
60
Description
- Graduated on both sides
- Tissue culture treated surface allows for effective cell adhesion and growth
- Comes with closed and filtered caps
- Leak-free
- Irradiation sterilized
- Pyrogen-free
- DNase-free, RNase-free

Specifications
Specifications
No. per Case | 60 |
No. per Pack | 10 |
Sterility | Sterile |
Culture Area | 6310 cm² |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction