Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Cell Culture Flasks with Vented Filter Cap, Tissue Culture Treated

Catalog No. p-8293005 Shop All Eppendorf North America Products
Change view
Click to view available options
Capacity (Metric):
7 mL
Culture Area:
26.9 cm2
Volume (Metric) Working:
7mL
No. per Case:
192
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Culture Area Volume (Metric) Working No. per Case
13-690-056 7 mL 26.9 cm2 7mL 192
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-690-056 Supplier Eppendorf™ Supplier No. 0030710126
This item is not returnable. View return policy

Preferred vessel for long term cultivation or large-scale expansion of cells, providing protection against contamination. Eppendorf™ Cell Culture Flasks with Vented Filter Cap, Tissue Culture Treated are sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free and non-cytotoxic.

  • Features ConvexAcces™ geometry for facilitated access to growth area and easier and safe cell treatment
  • Optimized gas exchange and protection against contamination through high efficiency air filter technology
  • Anti-rolling cap with corrugation facilitates cultivation steps when cap needs to be set aside
  • 100% in-line control for leakage-free flasks and maximum safety
  • Direct surface identification on flask and cap
  • Available in three sizes: T–25, T–75 and T–175 with TC treated surfaces
TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Capacity (Metric) 7 mL
Material Polystyrene
For Use With (Application)
  • Expansion and culture of adherent and suspension cells on TC treated or on non-treated polystyrene
  • Performance of cell-based assays and microscopic analysis
No. per Case 192
Product Type T-25 Cell culture flask
Surface Coating Tissue Culture Treated
Temperature Range (Metric) -86°C to +60°C
Autoclavable Not Autoclavable
Sterility Sterile SAL 10-6
Closure Type Vented filter cap
Culture Area 26.9 cm2
Volume (Metric) Working 7mL
Closure Size 27.2 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.