Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abbott Laboratories™ CELL-DYN Ruby
Click to view available options
Throughput:
84/hr
Description
- Uses MAPSS technology to enumerate and differentiate White Blood Cells and provide the performance needed for patient samples
- Uses optical laser light scatter analysis to provide both Red Blood Cell and Platelet counts
- Can process up to 84 CBCs and Differentials per hour
- MultiAngle Polarized Scatter Separation (MAPSS) laser technology provides enhanced first-pass efficiency, with accurate WBC enumeration and identification using 4 angles of light scatter on up to 10,000 cells per dilution
- Uses only 4 reagents including reticulocytes
- Sampling: Open and Closed
- Reagents: 3 and Retic
- Load up Capacity: 50 tubes with 10 per rack
- Maintenance Requirements: Auto-clean (15 min) daily and manual cleaning of syringes, shear valve, dil/sheath filter weekly
Specifications
Specifications
Sample Size | Open: 150 μL; Closed: 230 μL |
Throughput | 84/hr |
Depth (English) | 30.2 in. |
Height (English) | 19.6 in. |
Width (English) | 34 in. |
Weight (English) | 232 lb. |
Parameter | 20 and 2 Retic |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction