Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Cell Organization 3D Model Making Kit
SCI_ED

Catalog No. S436045 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S436045 6 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S436045 Supplier NewPath Learning™ Supplier No. 247738
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Cell Organization 3D Model Making Kit allows students to assemble models of cells, tissues, organs and organ systems to learn about animal cells, plant cells, lung tissue, plant leaf tissue, stomach, lung, leaf and digestive system.

  • Your students will first assemble the models of cells, tissues, organs and organ systems out of the colorful paper parts provided.
  • They then use these models to look at how they are related to each other.
  • Through the process they will learn about animal cells, plant cells, lung tissue, plant leaf tissue, stomach, lung, leaf and digestive system.
  • The Model Making Kit includes enough materials for students to assemble up to 5 paper models working cooperatively.
  • The comprehensive Teacher Resource Guide included provides illustrated readings, activities, key vocabulary and an assessment, along with teaching opportunities for students to Engage, Explore, Explain, Extend and Evaluate their understanding of the concepts presented.

Specifications

For Use With (Application) Model Assembly
Grade 6 to 10
Includes 5 Models, Teacher Resource Guide, Vinyl Pouch
Length (English) 12 in.
Material Paper
Weight (English) 2 lb.
Width (English) 9 in.
Product Type 3D Model
Grade Level Intermediate - Middle School, High School
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.