Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Cell Proliferation Kit, BioTrak™ ELISA Kit and Components

Catalog No. p-3667350
Click to view available options
Quantity:
130 μL
10 mL

Includes

The cell proliferation kit includes:
Labeling reagent, Nuclease, Anti-5-bromo-2'-deoxyuridine monoclonal antibody, Peroxidase anti-mouse IgG2a, Peroxidase anti-mouse IgG2a diluent, Substrate/intensifier

Cel proliferation Biotrak ELISA kit includes:
BrdU labelling reagent; Fixative; Blocking reagent; Peroxidase-labelled anti-BrdU; Antibody dilution solution; Wash buffer; Substrate solution

Cell proliferation labelling reagent and Anti-BrdU antibody are available separately

Provides a precise, fast and simple colorimetric assay to quantitate proliferating cells cultured in a 96 well microplate

  • Recommended cell number: 10e6-10e7 cells/mL (Cell proliferation kit) 10e2 cells/well-10e5 cells/well (Biotrak ELISA kit)
  • Required time: 2-3 hour post-labeling (labeling 1-2 hours)
  • Antibody and labeling reagent available separately for ELISA and ICC systems
  • Suitable for in vivo and in vitro tissue labeling in addition to cultured cells (ELISA format)
  • Non radioactive protocols provide sensitivity comparable to autoradiographic methods
  • Cell proliferation kit avoids the need to chemically denature DNA. Cellular morphology is preserved allowing multiple labeling experiments
  • 5-fluoro-2'-deoxyuridine is used to inhibit thymidilate synthetase increasing BrdU incorporation

When ordering the Cell Proliferation Kit (RPN20), you must also order DAB REAGENT OF RPN20/RPN1174 (RPNK8888)

TRUSTED_SUSTAINABILITY

Specifications

Includes 10mL of Aqueous 5-bromo-2'-deoxyuridine and 5-fluoro-2'-deoxyuridine
Quantity 10 mL
Content And Storage 2°C to 8°C
For Use With (Application) Immunocytochemistry
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.