Learn More
Thermo Scientific™ Reinforced Floor Option (Factory Installed)
Catalog No. 190777
Change view
Click to view available options
Installation:
Factory installed
Catalog No. Installation
19-077-7 Factory installed
1 options
Catalog No. 19-077-7 Supplier Thermo Scientific™ Supplier No. 190777
Only null left
Add to Cart
Add to Cart
For Use With: Model 3948 and 3962 Environmental Chamber/Incubator
Allow production of monolayer cell cultures in standard roller culture vessels with these Cell Roller System Accessories for Thermo Scientific™ Environmental Chambers and Incubators.
TRUSTED_SUSTAINABILITY
Specifications
Description Reinforced Floor with Removable Ramp (extends 23
For Use With (Equipment) Model 3948 and 3962 Environmental Chamber/Incubator
Lighting LED
Type Reinforced Floor Option
Installation Factory installed