Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cell Structure and Function CD-ROM

Click to view available options
Grade:
6 to 12
Description
- Virtual labs explore slide preparation, cell structure, cell membrane function, cell size, and diffusion
- Comprehensive tutorial uses illustrations, animations, and micrographs to give an overview of cell theory, animal and plant cell organelles, cell specialization, and the chemistry of cells
- Self-assessment feature tests and tracks student comprehension
- Detailed teacher's resource features illustrations for presentations and handouts, and helps create customized tests and lessons
Specifications
Specifications
Grade | 6 to 12 |
Certifications/Compliance | NSES |
Compatibility | Windows™ 98 or greater, Mac™ 8.6 or higher (classic mode) |
Category | Biology |
Format | CD-ROM |
For Use With (Application) | Explore slide preparation, cell structure, cell membrane function, cell size, and diffusion |
License | Single User |
Memory | 64MB |
Product Type | Cell Structure and Function |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction