Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell-Surface Vimentin (CSV) Mouse anti-Human, PE, Clone:84-1, Abnova™

Catalog No. 89333866 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-866 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-866 Supplier Abnova Corporation Supplier No. H00007431MP08
Add to Cart
Edge
Add to Cart

PE conjugated mouse monoclonal antibody recognizes human cell-surface vimentin (CSV).

This gene encodes a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein encoded by this gene is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a dominant, pulverulent cataract

Specifications

Antigen Cell-Surface Vimentin (CSV)
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 84-1
Conjugate PE
Description vimentin
Formulation PBS with no preservative; pH 7.4
Gene VIM
Gene Accession No. P08670
Gene Alias FLJ36605
Gene Symbols VIM
Host Species Mouse
Immunogen Human recombinant vimentin
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7431
Target Species Human
Content And Storage Store at 4°C in the dark. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.