Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CellBlox™ Plus Blocking Buffer

Catalog No. C001T02F01
Encompass_Preferred
Click to view available options
Quantity:
100 Tests
25 Tests
500 Tests

Invitrogen CellBlox Plus Blocking Buffer is formulated to block nonspecific binding of NovaFluor labels with cells.

Invitrogen CellBlox Plus Blocking Buffer is formulated to block nonspecific binding of NovaFluor labels with cells. These nonspecific interactions can result in higher background labeling. CellBlox Plus Blocking Buffer is a non-antibody blocking solution and should be used whenever a NovaFluor dye is used for labeling of any cell type. CellBlox Plus Blocking Buffer has enhanced blocking capabilities compared to our original CellBlox Blocking Buffer (Figure 1).

CellBlox Plus Blocking Buffer is also recommended for use with cyanine-based dyes or cyanine-based tandem dyes to block non-specific interactions with monocytes and macrophages to minimize background labeling (Figure 2).

Features of CellBlox Plus Blocking Buffer include:
• Compatible with all fluorophores and with LIVE/DEAD Fixable dead cell stains
• Can be used with any fluorophore-antibody conjugate as high-performance monocyte and macrophage blocking solution
• Compatible with other blocking reagents, such as Fc Block, blocking proteins, Brilliant Stain Buffer, and Super Bright Complete Staining Buffer
• Not required when labeling antibody-capture beads, such as UltraComp eBeads Plus Compensation Beads

Use of CellBlox Plus Blocking Buffer requires minimal change to most flow cytometry staining protocols. Simply add CellBlox Plus Blocking Buffer to an antibody cocktail mixture prior to labeling cells. Specifically, add 5 μL CellBlox Plus Blocking Buffer for every stained sample to be labeled with the antibody cocktail mixture, with 100 μL as a final staining volume.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Flow Cytometry
For Use With (Equipment) Flow Cytometer
Physical Form Liquid
Product Line eBioscience
Quantity 25 Tests
Solution Type Buffer
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.