Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One CELLCOAT™ Collagen Type I Coated Cell Culture Flask
Shop All Greiner Bio-One Products
Click to view available options
Culture Area:
75 cm2
Volume (Metric) Working:
15 to 38 mL
Description
Poly-D-lysine (PDL) are synthetic molecules which increase cell adhesion to vessels by altering the net charge of the surface without stimulating biological activity.
- Coated with Poly-D-Lysine synthetic protein (75-150 kDa)
- For promotion of cell adhesion, proliferation and growth of transfected cell lines, neuronal cell lines, primary neurons and glial cells
- Ready-to-use products
- Includes lid with condensation rings
Specifications
Specifications
| Closure Color | Red |
| Closure Material | HDPE |
| Capacity (Metric) | 250 mL |
| Material | Polystyrene |
| Neck Style | Canted |
| No. per Case | 50 |
| No. per Pack | 5 |
| Surface Coating | Collagen Type I Protein |
| Graduated | Yes |
| Closure Type | Filter Screw Cap |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction