Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One CELLCOAT™ 96-well, Poly-L Lysine-Treated, Flat-Bottom Microplate
Shop All Greiner Bio-One Products

Click to view available options
Color:
Black, Clear Bottom
Description
Poly-L-Lysine (PLL) are synthetic molecules which increase cell adhesion to vessels by altering the net charge of the surface without stimulating biological activity.
- Coated with Poly-L-Lysine synthetic protein (30-70 kDa)
- For promotion of cell adhesion, proliferation and growth of transfected cell lines, neuronal cell lines, primary neurons and glial cells
- Ready-to-use products
- Includes lid with condensation rings
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Content And Storage | Room Temperature |
| Culture Area | 0.34 cm2 |
| Array | 8 x 12 |
| Color | Black, Clear Bottom |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | Poly-L-Lysine |
| Show More |
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction