Learn More
Corning™ CellCube™ Modules, With Aseptic Connectors
GREENER_CHOICE
Catalog No. p-7221304
Click to view available options
Tray Layers:
10-Layer
100-Layer
25-Layer
This item is not returnable. View return policy
This item is not returnable. View return policy
Provides a fast, simple, and compact method for the mass culture of attachment-dependent cells.
  • Uses a tissue culture-treated growth surface for cell attachment, and continually perfuses the cells with fresh medium for increased cell productivity
  • The CellCube system provides an environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen with low differential gradients across all cells within the modules.
  • The CellCube modules provide a traditional tissue culture-treated surface or Corning CellBIND™ surface for the growth of attachment dependent cells.
TRUSTED_SUSTAINABILITY
Specifications
Includes Growth Surface and Aseptic Connectors
No. per Case 2/Cs.
No. per Pack 1/Pk.
Sterility Sterile
Culture Area 8500 cm²
Tray Layers 10-Layer
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.