Learn More
Invitrogen™ Celleste™ 3D Visualization Module, network license (includes one seat)
Catalog No. 12563702
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
12-563-702 1 each
1 options
Catalog No. 12-563-702 Supplier Invitrogen™ Supplier No. AMEP4917
This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or www.fishersci.com/cellanalysisdemo to request a demonstration customized to your application.
The Celleste 5.0 Image Analysis package easily transforms images into quantitative data in a streamlined workflow. The Celleste Image Analysis 3D Visualization Module for Network License helps enhance the 3D object visualization using a range of powerful tools following 3D deconvolution.
The Celleste 5.0 Image Analysis package easily transforms images into quantitative data in a streamlined workflow. The Celleste Image Analysis 3D Visualization Module for Network License helps to enhance the 3D object visualization using a range of powerful tools after 3D deconvolution. Coupled with the powerful image acquisition capabilities of the EVOS microscopes, Celleste 5 software allows you to seamlessly capture, process, measure, analyze and share images and data. A Network Server License is required for use with this module.
Celleste 5.0 Image Analysis Software Network License [1] is a full-feature image analysis package designed for a range of biological applications, from image adjustments and processing with manual and automatic measurements over multiple channels, to segmentation and classification tools that help you transform images into quantitative data in a streamlined and customizable workflow. The Celleste 5.0 Image Analysis Network Server License is designed for environments where the use of multiple copies is desired. The Network Server License comes with one seat and additional Soft License Seats [2] can be added at any time. Optional modules for 2D and 3D deconvolution, visualization, analysis and GPU-powered processing allow you to customize the capabilities according to your needs. The Celleste 3D Visualization Module for Network License helps to enhance the 3D object visualization using a range of powerful tools following 3D deconvolution.Coupled with the powerful image acquisition capabilities of the EVOS microscopes [3], Celleste 5 software allows you to seamlessly capture, process, measure, analyze and share images and data.Note that Celleste 5.0 Image Analysis Network Server License is required for use with any of the modules.Celleste 5.0 software with the Celleste 3D Visualization Module for Network License offers you these important advantages:• Streamlined workflow tools that make it easy to visualize and edit images• Powerful image analysis capabilities for segmentation, classification, and quantification of single images or a batch of images• Background and transparency adjustment• Choice of different projection types that best suit your images• Creation and enhancement of 3D movies by key frame annotations, object counts, and intensity adjustmentsMore about the deconvolution modulesDeconvolution is an algorithm-based process used to restore systematic loss of image contrast (blur) that results from the image point spread function (PSF). The powerful 2D and 3D restoration algorithms available in the Celleste 5 modules allow you to deconvolve your images to enhance contrast and quality, or to visualize them in Z, over time and by channel, then analyze the resulting image using an expanded tool set.[1] Celleste 5.0 Image Analysis Network Server License [AMEP4868][2] Celleste Image Analysis Network Seat [AMEP4869]Explore the entire EVOS line of imaging systems, accessories, and offers [3] [www.thermofisher.com/EVOS]For Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Celleste
For Use With (Equipment) EVOS M5000, EVOSM7000 and Other Imaging Systems
Quantity 1 each