Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CellEvent™ Caspase-3/7 Detection Reagents

Catalog No. C10723
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Vial
1 x 100 μL Vial
1 x 25 μL Vial
2 Vials (1 Green, 1 Red)
5 Vials
Color:
Green
Red
Red, Green
Excitation/Emission:
∼502/530 nm
∼590/610 nm
∼590/610 nm, ∼502/530 nm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color Excitation/Emission
C10723 1 x 25 μL Vial Green ∼502/530 nm
C10430 1 Vial Red ∼590/610 nm
C10431 5 Vials Red ∼590/610 nm
C10432 1 Vial Green ∼502/530 nm
C10433 5 Vials Green ∼502/530 nm
C10434 2 Vials (1 Green, 1 Red) Red, Green ∼590/610 nm, ∼502/530 nm
C10423 1 x 100 μL Vial Green ∼502/530 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. C10723 Supplier Invitrogen™ Supplier No. C10723
Add to Cart
Edge
Add to Cart

CellEvent Caspase-3/7 detection reagents are novel fluorogenic substrates for apoptosis detection. These substrates detect activated caspase-3/7 in live cells that can be fixed and multiplexed with antibody detection workflows.

CellEvent Caspase-3/7 Green and CellEvent Caspase-3/7 Red detection reagents are novel fluorogenic substrates for activated caspase-3/7 in live cells that can be fixed and multiplexed with antibody detection workflows such as immunofluorescence (IF) and immunocytochemistry (ICC). Activation of caspase-3 is an essential event during apoptosis, making these optimized reagents for analysis of apoptotic cells. CellEvent Caspase-3/7 Red Detection Reagent can also be used to detect apoptosis in GFP expressing cells.

Features of these indicators include:

  • Optimized caspase-3/7 substrate for apoptosis analysis
  • Easy to use—add and read; no cell lysis required
  • Use for time course measurements; easily select the time point of interest
  • Compatible with both live cell fluorescence-imaging and formaldehyde-based fixation methods
  • Multiplex enabled—combine with other fluorescent reagents to confirm apoptosis in the same cell or cell population

CellEvent™ Caspase-3/7 Green Detection Reagent: Absorption/emission maxima of ∼502/530 nm. Detectable with FITC/GFP traditional filter set.

CellEvent™ Caspase-3/7 Red Detection Reagent: Absorption/emission maxima of ∼590/610 nm. Detectable with traditional Texas red filter set.

CellEvent™ Caspase-3/7 Red Detection Reagent and CellEvent™ Caspase-3/7 Green is provided as Dry down powder. Just add 100μL of PBS to dry down powder to generate 100X Stock solution. Dilute this 100X stock solution 1:100 in complete media and stain cells for 30-60 minutes and analyze with fluorescence microscope.

CellEvent™ Caspase-3/7 Green Detection Reagent is also provided as as already prepared DMSO solution. DMSO solution is 400X stock solution, which is ready to use. Just dilute in growth media 1:400 and stain the cells for 30 to 60 minutes.

To prevent the apoptotic cells from washing away, no wash is needed at the end of staning. Stained cells can be analyzed live, or can be fixed with 4% Paraformaldehyde solution such as Image-IT™ Paraformaldehyde 4% in 0.1M Phosphate Buffer pH 7.4, Cat. No. I28800. These fixed cells can be further processed for ICC workflow.

Substrate specifics

CellEvent Caspase-3/7 Detection Reagents are a four-amino acid peptide (DEVD) conjugated to a nucleic acid-binding dye. The DEVD peptide sequence is a cleavage site for caspase-3/7, and the conjugated dye is non-fluorescent until cleaved from the peptide and bound to DNA.

Principles of action

CellEvent Caspase-3/7 Green Detection Reagent is intrinsically non-fluorescent as the DEVD peptide inhibits the ability of the dye to bind to DNA. However, after activation of caspase-3/7 in apoptotic cells, the DEVD peptide is cleaved, enabling the dye to bind to DNA and produce a bright, fluorogenic response.

Simple three-step protocol

To use CellEvent Caspase 3/7 Detection Reagent, simply add it to cells, incubate 30–60 minutes, and visualize. Apoptotic cells with activated caspase-3/7 will have bright green or red nuclei, while cells without activated caspase 3/7 will have minimal fluorescent signal.

Assay versatility allows for live cell detection and fixation

This robust assay enables the examination of caspase-3/7 activation in live cells. Furthermore, since wash steps are not necessary for detection, fragile apoptotic cells typically lost during wash steps are preserved. Importantly, the fluorescent signal from CellEvent Caspase 3/7 Detection Reagent can survive fixation and permeabilization. This allows for the flexibility to perform end-point assays and probe for other proteins of interest using immunocytochemistry.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 x 25 μL Vial
Product Type Caspase 3/7 Reagent
Content And Storage 1 x 25 μL 2 mM reagent in DMSO. Store at ≤-20°C, desiccate, and protect from light.
Excitation/Emission ∼502/530 nm
Color Green
Shipping Condition Room Temperature
Product Line CellEvent
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, High Content Instrument
Label or Dye CellEvent™ Caspase 3/7 Green
Label Type Other Label(s) or Dye(s)
Form 2 mM in DMSO
Show More Show Less
I want to use your CellEvent Caspase-3/7 Green detection reagent for studying apoptosis in live cells, but for 96-well microplate instead of microscopy. Will it work? Can I multiplex it with a cell vitality dye?

Yes, on both accounts. We have demonstrated that it works for microplates, and we have multiplexed the dye with PrestoBlue Cell Viability Reagent. Some microplates may be less sensitive than microscopic analysis, so sensitivity may not be the same for the CellEvent reagent.

How long can I measure the Cell Event Caspase-3/7 Detection Reagents (Cat. Nos. C10432, C10433, C10434, C10423, C10723, R37111) signal for the kinetic assay?

We have tested Caspase 3/7 Green Detection Reagents (Cat. Nos. C10432, C10433, C10434, C10423, C10723, R37111) for up to 48 hours and observed no toxicity or stability issues with the reagent. If the samples are fixed at the end of the time course the signal will be retained for days. For live cells, apoptotic cells will round up and float away before the loss of fluorescence signal from CellEvent.

The following reference reports a visible signal for up to 72 hrs:

Sambi M, Samuel V, Qorri B, Haq S, Burov SV, Markvicheva E, Harless W, Szewczuk MR. A Triple Combination of Metformin, Acetylsalicylic Acid, and Oseltamivir Phosphate Impacts Tumour Spheroid Viability and Upends Chemoresistance in Triple-Negative Breast Cancer. Drug Des Devel Ther. 2020 May 25;14:1995-2019. doi: 10.2147/DDDT.S242514. PMID: 32546966; PMCID: PMC7260544.

Can I use CellEvent Caspase-3/7 Green Detection Reagent with fixed samples?

No, it must be applied to live cell populations.

With the CellEvent Caspase-3/7 Green Detection Reagent, may I stain cells with nucleic acid stains?

Yes, you may use SYTOX AADvanced Dead Cell Stain that is included in the CellEvent Caspase-3/7 Green Ready Flow Reagent (Cat. No. R37167) and in the CellEvent Caspase-3/7 Green Flow Cytometry Assay Kit (Cat. No. C10427, C10740). You may also try using other nucleic acid stains in the red or far red channels for staining cells.

With the CellEvent Caspase-3/7 Green Detection Reagent, may I multiplex with fluorescent immunolabeling?

Yes. You can apply surface labels or other reagents. We recommend applying the surface labels or other reagents first.

Is the CellEvent Caspase-3/7 Green Detection Reagent fixable?

The CellEvent Caspase-3/7 Green Detection Reagent is not covalently attached to any cellular component and may be lost upon fixation. For flow cytometry applications, after applying the reagent, you should examine without washing or fixation. For imaging applications, the CellEvent product signal may be retained after fixation in 3.7% formaldehdye for 15 min.

Can I fix my CellEvent Caspase 3/7 - labeled cells?

For imaging applications, we recommend fixing with 3.7% formaldehyde at room temperature for 15 mins. After fixation, cells can be imaged for up to 24 hrs, but the signal will gradually degrade over time. For flow cytometry applications, fixation is not recommended.

I wish to measure cell proliferation in combination with apoptosis in a microplate assay. Do you have any product recommendations?

It is possible to combine PrestoBlue reagent (for cell proliferation) with the CellEvent Caspase 3/7 Green Detection Reagent (for apoptosis detection) in the same sample.

What are the advantages of flow cytometry?

-Measures data from single cells.
-Data are obtained for a large number of cells, generating a rich statistical analysis of cell populations.
-Because single cells are measured, it will reveal heterogeneity within a population.
-With the ability to multiplex, small sub-populations can be identified.
-Thousands of cells can be analyzed rapidly.
-It is ideally suited for blood samples and other cells in suspension.
-Data can be re-analyzed multiple times after acquisition.
-Flow cytometry files (FCS) can be archived.

What kinds of applications can I run on a flow cytometer?

There are several applications, some of which include immunophenotyping, cell cycle analysis, apoptosis assays such as annexin V staining, CellEvent Caspase-3/7 assay, and TUNEL assay, cell viability, proliferation assays such as CellTrace assay and Click-iT EdU assay, measurements of mitochondrial potential with MitoProbe assays, and cell counting using counting beads.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.