Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CellInsight™ CX5 High Content Screening (HCS) Platform

Catalog No. cx51110
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CX51110 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CX51110 Supplier Thermo Scientific™ Supplier No. CX51110

This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or www.fishersci.com/cellanalysisdemo to request a demonstration customized to your application.

Rapidly perform high content imaging assays with this affordable, compact HCS platform.

Rapidly perform high content imaging assays with the affordable, compact Thermo Scientific™ CellInsight™ CX5 High Content Screening (HCS) Platform.

The CellInsight CX5 High Content Screening (HCS) Platform is a fast, simple-to operate, automated cellular imaging and analysis benchtop platform designed for quantitative imaging. For the researcher looking to gain the advantage of quantitative cell analysis for the first time or for those who are looking to increase scale and capacity, the CellInsight CX5 HCS Platform offers a costs effective, but powerful way to quickly automate quantitative cellular imaging.

Highlights:

  • Flexible – designed to work with or without fluorescent labeling, the new CellInsight CX5 HCS Platform uses transmitted light allowing users to explore more cell biology without the restriction of fluorescent dyes or to use the full capabilities of a five-color light engine
  • Affordable – automated high content imaging for all budgets
  • Easy to Use – quickly build and optimize image-based assays with included Thermo Scientific™HCS Studio™ 2.0 Cell Analysis Software
  • Performance – scan and analyze a 96-well plate in under four minutes
  • Quality – high-quality imaging and virtually maintenance-free operation
  • Compact – fits on a bench and is less than 2 ft. on all sides

Capabilities:

  • Make individual cell, subcellular and multicellular measurements
  • Save time by automatically collecting only the images and data you need to be statistically relevant
  • Make multiple measurements per object, over time if desired
  • Measure subpopulations and reject undesired objects automatically

Easy to Use Applications:

  • Comprehensively pre-validated and user configurable image analysis without scripting or programming
  • Exploratory assay development tools and specific biology optimized tools
  • Define biological thresholds for outputs based on control populations
  • Ability to create multi-parameter gates from individual output measurements

Hardware and Software:

  • HCS Studio 2.0 Cell Analysis Software – the premier high content instrument control, visualization and analysis program for novice to advanced users featuring image/data portability
  • Ultra-stable, long-life light source reduces variability, increases data quality, and decreases operating costs
  • Fluorescence options which include near-IR wavelengths
  • Proprietary Thermo Scientific™ AccuFocus System is fast, yet flexible, imaging cells with high-fidelity every time
  • Unique intelligent acQuisition (iQ) real time analysis feedback to optimize image collection and data quality
  • Compatible with SBS standard microplates (6-1536 well) and slides

Compatible with:
Optional Automation – Thermo Scientific™ Orbitor™ RS Plate Mover and other Thermo Scientific™ Laboratory Automation Solutions; Optional Modules - Thermo Scientific™ High Content Modules

TRUSTED_SUSTAINABILITY

Specifications

Product Line CellInsight
Description CeLLInsight™ CX5 High Content Screening Platform with HCS Studio Cell Analysis Software and Store Express
For Use With (Equipment) Thermo Scientific™ Orbitor™ RS Plate Mover
Color Green, Blue, Near-Infrared, Far-Red, Red
Detection Method Brightfield, Fluorescence
Format Slide(s), 96-well plate, 384-well plate, 6-well plate, 1536-well plate, 8 wells, 24-well plate
High-throughput Compatibility High-throughput Compatible, Multiplexing, Automated Protocols
Wavelength 740⁄809, 560⁄607, 485⁄521, 386⁄440, 650⁄694
Objectives Additional Objectives Available, Low and High NA, Single Position 2x-20x
Optics CCD Camera, LED, solid-state 5-color light engine
Data Outputs JPG Files, TIFF Files, PNG Files, C01, BMP, BIN, PZFX, ICE, GIF Files, DIB, CSV, FCS 3.1, XLSX
Display 24in High-resolution widescreen monitor
Memory 8GB RAM
Operating System Windows 7
PC Windows 7™ 64-bit PC with quad core 3.7GHz processor, 8GB RAM, and 500GB Hard Drive
Software Compatibility Thermo Scientific™ Store™ Image and Database Management Software, Thermo Scientific™ HCS Studio™ Cell Analysis Software
Operating Environment Permissible ambient temperature range during operation : + 18 deg C to + 25 deg C, Permissible relative humidity during operation (noncondensing): > 75% at 30 deg C
Dimensions 41.4 cm (16.3 in) (W) x 37.2 cm (14.6 in) (H) x 59.4 cm (23.4 in) (D)
Weight (Metric) 45 kg
Quantity 1 each
Voltage 120V
Wattage 300 W
Show More Show Less
Who do I contact if I have issues with my CellInsight systems or ArrayScan System?

Contact Instrument Customer Services for any issues regarding the CellInsight systems and ArrayScan System. They can be reached at instrumentservices@thermofisher.com.

For high-content analysis (HCA)/high-content screening (HCS), what are the main differences between the CellInsight CX5, CX7, and ArrayScan systems?

The basic differences are in the number of channels for excitation (5 versus 7), the ability to image in brightfield, widefield, and confocal, and the number of positions available on the objective turret. The CellInsight CX5 System is the most basic HCS system with illumination in 5 channels, imaging in brightfield and widefield, and a 1-position objective turret. The CellInsight CX7 System and ArrayScan System provide illumination in 7 channels, imaging in brightfield, widefield, and confocal, and either a 3-position (CX7) or 4-position objective turret (ArrayScan System). The ArrayScan System has the added option of live-cell imaging. For more information, go here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cellular-imaging/high-content-screening.html).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.