Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellosolve™ 2-Ethoxyethanol, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187771
Click to view available options
Quantity:
5 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Ethoxyethanol is an organic compound, widely used as a component or solvent for nitrocellulose and in a variety of dyes, inks, cleaning agents, resins, paints, varnishes, etc. It is also used for increasing the stability of emulsions.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line Cellosolve
Ignition Point 460°F
CAS 110-80-5
Vapor Pressure 3.8 mmHg (20°C)
Vapor Density 3.1 (vs. air)
Density 0.93 g/mL (at 25°C)
Flash Point 42°C
Physical Form Neat
Linear Formula C2H5 OCH2 CH2 OH
MDL Number MFCD00002869
Melting Point -90°C
Boiling Point 135°C
Synonym Cellosolve(R); Ethyl glycol; Ethylene glycol monoethyl ether
Type Analytical Standard
Quantity 5 mL
Formula Weight 90.12
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less

Cellosolve is a registered trademark of Union Carbide

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.