Learn More
Cellpath CellSafe+ Blue Biopsy Capsules
SDP
Catalog No. p-4908705
Click to view available options
Quantity:
100/Pk.
1000/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy
Offers high security and easy to use alternative to traditional biopsy handling techniques such as tissue wrap and foam pads which are proving to be unsatisfactory and time consuming. CellPath CellSafe+ Blue Biopsy Capsules features new blue mesh that provides better contrast between the biopsy and the CellSafe.
  • Features two hinged interlocking frames with an integral nylon mesh
  • Adjustable to biopsy specimen size
  • Hinged in either direction and closed to encapsulate the tissue
  • New Blue Mesh provides better contrast between the biopsy and the CellSafe
  • X–Ray transparent mesh for breast work
  • Extra fine mesh provides up to 20 times less carry–over of solvents than foam pads
  • Zero Tissue damage or artefacts associated with foam pads
  • Substantial time savings compared to folding biopsy in tissue paper
Specifications
Quantity 100/Pk.
Color Blue
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.