Learn More
Cellpath CellSafe+ White Biopsy Capsules
Click to view available options
Quantity:
100/Pk.
1000/Pk.
Description
- Features two hinged interlocking frames with an integral nylon mesh
- Adjustable to biopsy specimen size
- Hinged in either direction and closed to encapsulate the tissue
- X–Ray transparent mesh for breast work
- Extra fine mesh provides up to 20 times less carry–over of solvents than foam pads
- Zero Tissue damage or artefacts associated with foam pads
- Substantial time savings compared to folding biopsy in tissue paper
Specifications
Specifications
| Quantity | 100/Pk. |
| Color | White |