Learn More
Cellpath System II Hex™ Tissue Processing Cassettes
Click to view available options
Color:
Blue
Brown
Dark Blue
Dark Green
Gray
Green
Lavender
Orange
Peach
Pink
Red
Tan
Turquoise
Yellow
Description
- Manufactured from high–grade virgin acetyl resin
- Hexagonal perforations arranged in honeycombed structure to better utilize the available surface area
- Unique arrangement increases open area thereby increasing reagent flow through and improving specimen staining and analysis time
- Extra thick side walls increases rigidity and stability for high quality sectioning
- Available in 15 stock colors which color code different types of workload or applications
Specifications
Specifications
Quantity | 1000/Pk. |
Color | Blue |
Material | Acetyl Resin |
Product Type | Tissue Processing Cassette |