Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One CELLSTAR 384-well, Cell Culture-Treated, Flat-Bottom Microplate

Catalog No. 07001037 Shop All Greiner Bio-One Products
Click to view available options
Packaging:
20/Pk., 120/Cs.
8/Pk., 32/Cs.

Color: White; Sterility: Sterile; Lid: With Lid

  • Made from polystyrene
  • Tissue-culture treated
  • Solid or μClear™ flat bottom
  • With lid

Service Plan

No

Warranty and Services

No

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid With Lid
Array 16 x 24
Packaging 20/Pk., 120/Cs.
Color White
Product Type Cell Culture Plate
Material Polystyrene
No. of Wells 384
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.