Learn More
Greiner Bio-One CELLSTAR Cell-Repellent Surface Cell Culture Flasks with Screw Cap
Catalog No. 07000667 Shop All Greiner Bio-One Products
Click to view available options
Culture Area:
175 cm2
25 cm2
75 cm2
Volume (Metric) Working:
50mL
250mL
650mL
Ideal for suspension, spheroid cultures and stem cell aggregate formation. Greiner Bio-One CELLSTAR Cell-Repellent Surface Cell Culture Flasks with Screw Cap effectively inhibit cell adhesion.
  • Made of polystyrene
  • Used for applications such as Gel-based 3D cultures and Magnetic 3D cell culturing (Nano3D technology)
  • Cell-repellent surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments
  • 660980: Flat flask design and 661980: High flask design
TRUSTED_SUSTAINABILITY
Specifications
Closure Color White
Description High flask design
Material Polystyrene
Surface Coating Cell Repellent
Closure Type Screw Cap
Culture Area 175 cm2
Volume (Metric) Working 650mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.