Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One CELLSTAR™ Small Volume™ 384 Well Flat Bottom Cell Culture Plate

Improves cell adhesion. Greiner Bio-One CELLSTAR™ Small Volume™ 384 Well Flat Bottom Cell Culture Plate is surface treated to enhance cell growth

Manufacturer:  Greiner Bio-One 788073

 View more versions of this product

Catalog No. 07-000-921


Add to cart

Description

Description

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic, non-cytotoxic
  • Compatible with automated systems
  • Stackable
  • Alphanumeric well coding
  • Growth area per well: 2.7mm²
  • Working volume per well:
Specifications

Specifications

Tissue-culture Treated
White
Polypropylene
Tissue-Culture Treated
With Lid
28μL
Sterile
Solid
384 well
384
−20° to 60°C
10/ bag
Microplate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit