Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CellTrace™ Violet Cell Proliferation Kit, for flow cytometry

Catalog No. p-7075734
Encompass_Preferred
Change view
Click to view available options
Quantity:
180 Assays
20 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34557 180 Assays
50-112-1516 20 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C34557 Supplier Invitrogen™ Supplier No. C34557
Only null left
Add to Cart
Edge
Add to Cart

CellTrace™ Violet Cell Proliferation Kit, for flow cytometry

CellTrace™ Violet Cell Proliferation Kit is used for in vitro and in vivo labeling of cells to trace multiple generations using dye dilution by flow cytometry.

  • Superior performance—bright, single-peak staining enables visualization of multiple generations
  • Long-term signal stability—well-retained in cells for several days post stain
  • Versatile—multiple colors available to easily combine with antibodies or markers of cell function, such as GFP
  • Simple, robust staining protocol

    View a selection guide for all CellTrace™ Cell Proliferation Kits for flow cytometry.

    Superior fluorescent staining
    Successful proliferation analysis by dye dilution (see figure below) requires an extremely bright dye to distinguish fluorescently labeled cells from auto-fluorescence after several cell divisions. The intense fluorescent staining provided by CellTrace™ Violet dye enables the visualization of ten or more generations of proliferating cells before the signal is overwhelmed by intrinsic cellular auto-fluorescence. Consistent, homogeneous staining results in very little fluorescence variation between cells in a population, so distinct generations can be seen without any requirement for complex modeling software.

    Long-term signal retention
    Unlike stains that label the lipid membrane of cells, CellTrace™ Violet stain easily crosses the plasma membrane and covalently binds inside cells where the stable, well-retained fluorescent dye provides a consistent signal, even after several days in a cell culture environment. CellTrace™ Violet dye binds covalently to all free amines on the surface and inside of cells and shows little cytotoxicity, with minimal observed effect on the proliferative ability or biology of cells.

    Easy multiplexing with other fluorophores
    The violet excitation and narrow emission of CellTrace™ Violet dye make it ideal for multiplexing due to the limited spectral overlap with other common dyes (Alexa Fluor™ 488, FITC, and RPE) and fluorescent proteins (Green Fluorescent Protein (GFP) and mCherry) (see Fluorescence Spectra for CellTrace™ Violet stain below).

    Simple, robust staining protocol
    The CellTrace™ Violet Cell Proliferation Kit contains convenient single-use vials of dry dye to permit small-scale experiments without preparing excess quantities of dye. A stock solution is prepared by dissolving the contents of a vial in anhydrous DMSO prior to use. To stain 1 mL of cells in protein-free medium, 1μL of this stock solution is typically used. Cells should be stained for 20 minutes at room temperature with gentle agitation. A brief wash with complete medium will then quench any dye remaining in solution.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Contains 9 vials of CellTrace™ Violet (lyophilized powder) and 1 vial of DMSO (500 μL). Store in freezer (-5 to -30°C) and protect from light.
    Detection Method Fluorescence
    For Use With (Application) Proliferation Assay
    For Use With (Equipment) Flow Cytometer
    Product Type Cell Proliferation Kit
    Dye Type Other Labels or Dyes
    Emission 405/450
    Form Lyophilized
    Format Tube(s)
    Product Line CellTrace
    Quantity 180 Assays
    Shipping Condition Room Temperature
    Solubility DMSO (Dimethylsulfoxide)
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.