Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellular Apoptosis Susceptibility (CSE1L) Monoclonal Antibody (OTI2C5), TrueMAB™, OriGene
SDP

Catalog No. 502543774
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3774 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3774 Supplier OriGene Supplier No. CF812194
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cellular Apoptosis Susceptibility (CSE1L) Monoclonal antibody specifically detects Cellular Apoptosis Susceptibility (CSE1L) in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Cellular Apoptosis Susceptibility (CSE1L)
Applications Western Blot
Classification Monoclonal
Clone OTI2C5
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene Numbl
Gene Accession No. A1L1I3, O08919, Q9Y6R0
Gene Alias 2610100P18Rik; AA407533; Capts; CAS; Cellular apoptosis susceptibility protein; chromosome segregation 1 like; chromosome segregation 1-like (S. cerevisiae); chromosome segregation 1-like protein; CSE1; CSE1 chromosome segregation 1 like; CSE1 chromosome segregation 1-like; CSE1 chromosome segregation 1-like (yeast); CSE1L; Exp2; exportin-2; Importin-alpha re-exporter; MGC117283; MGC130036; MGC130037; XPO2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-225 of human NUMBL (NP_004747) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18223, 292732, 9253
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.