Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Cellulase (T. reesei)

Catalog No. 89939182
Click to view available options
Quantity:
250 mg

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Activity
Name Cellulase (T. reesei)
Preparation Method Native protein chromatographically purified from Trichoderma reesei ATCC#26921
Quantity 250 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name CELLULASE
Assays Cellulase Assay
Biological Activity ≥45 units per mg dry weight. One Unit releases 0.01 milligrams of glucose per hour from microcrystalline cellulose at 37°C, pH 5.0.
Species Fungi
Protein Tag None
Expression System Native protein chromatographically purified from Trichoderma reesei ATCC#26921
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.