Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellulose microcrystalline, average particle size 50 μm

Catalog No. p-3586413
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic drum

CAS: 9004-34-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-34-6
MDL Number MFCD00081512

Specifications

Color White
pH 5 to 7
Physical Form Micro Fine Crystalline Powder
Linear Formula (C12H20O10)n
Solubility Information Solubility in water: insoluble.
Quantity 250 g
Infrared Spectrum Authentic
Loss on Drying 1.5% max.
Packaging Plastic Bottle
Particle Size 250 Micron <1% min., 75 Micron <30% min., 32 Micron >50% min.
Residue after Ignition 0.05% max.
Chemical Name or Material Cellulose microcrystalline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.