Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CellVation™ Cryopreservation Medium

Catalog No. p-8015219 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
60 mL
1 L

Catalog No. MP920300M2

Supplier: MP Biomedicals™ 0920300M2

Only null left
Add to Cart
Add to Cart

Recommended for the preservation of stem cells, hepatocytes, tissue samples, and a wide variety of cell types and established cell lines

CellVation is a cryopreservation medium designed for optimal cell recovery and convenience. It does not contain DMSO or serum, and is scientifically engineered for serum-free cultures, as well as, ideally suited for all types of serum. It has also been shown to preserve the potential of stem cells for differentiation.

TRUSTED_SUSTAINABILITY

Specifications

Form Solution
Product Type Cryopreservation Media
For Use With (Application) Cell counting, viability and cryopreservation
Quantity 60 mL
Certifications/Compliance Mycoplasma, Bacteria, Fungi, Yeast and Mold free. Certified U.S. origin.
Endotoxin Level <0.5 EU/mL
pH 6.5 to 7.5
Content And Storage Store at 4°C
Description CellVation Cryopreservation Media

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.