Learn More
Invitrogen™ CELSR1 Polyclonal Antibody

Description
Immunogen sequence: PTSFRLQILN NYLQFEVSHG PSDVESVMLS GLRVTDGEWH HLLIELKNVK EDSEMKHLVT MTLDYGMDQN KADIGGMLPG LTVRSVVV Highest antigen sequence indentity to the following orthologs: Rat - 60%, Mouse - 58%.
Specifications
Specifications
| Antigen | CELSR1 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CELSR1 |
| Gene Accession No. | Q9NYQ6 |
| Gene Alias | ADGRC1; adhesion G protein-coupled receptor C1; cadherin EGF LAG seven-pass G-type receptor 1; Cadherin family member 9; cadherin, EGF LAG seven-pass G-type receptor 1 (flamingo homolog, Drosophila); CDHF9; CELSR1; Flamingo homolog 2; FMI2; hFmi2; ME2; protocadherin flamingo 2 |
| Gene Symbols | CELSR1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.