Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CELSR2 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298104MFV610
Description
CELSR2 Polyclonal antibody specifically detects CELSR2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
CELSR2 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
cadherin EGF LAG seven-pass G-type receptor 2, Cadherin family member 10, cadherin, EGF LAG seven-pass G-type receptor 2 (flamingo homolog, Drosophila), CDHF10FLJ34118, EGFL2FLJ45143, EGF-like protein 2, EGF-like-domain, multiple 2, epidermal growth factor-like 2, Epidermal growth factor-like protein 2, Flamingo homolog 3, Flamingo1, FLJ45845, KIAA0279FLJ42737, MEGF3cadherin, EGF LAG seven-pass G-type receptor 2, flamingo (Drosophila) homolog, Multiple EGF-like domains protein 3, multiple epidermal growth factor-like domains 3, Multiple epidermal growth factor-like domains protein 3 | |
Produced in rabbits immunized with E. coli-derived Human CELSR2 fragment. | |
0.1 mL | |
GPCR, Growth and Development, Neuronal Cell Markers | |
1952 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction