Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CELSR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533423
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533423 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533423 Supplier Invitrogen™ Supplier No. PA533423
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset, Mouse, Rat, Bovine, Dog, Hamster, Elephant, Panda, Horse, Rabbit (100%) Opossum (94%).

The protein encoded by this gene is a member of the flamingo subfamily, part of the cadherin superfamily. The flamingo subfamily consists of nonclassic-type cadherins; a subpopulation that does not interact with catenins. The flamingo cadherins are located at the plasma membrane and have nine cadherin domains, seven epidermal growth factor-like repeats and two laminin A G-type repeats in their ectodomain. They also have seven transmembrane domains, a characteristic unique to this subfamily. It is postulated that these proteins are receptors involved in contact-mediated communication, with cadherin domains acting as homophilic binding regions and the EGF-like domains involved in cell adhesion and receptor-ligand interactions. The specific function of this particular member has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CELSR2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene CELSR2
Gene Accession No. Q9HCU4, Q9QYP2, Q9R0M0
Gene Alias ADGRC2; adhesion G protein-coupled receptor C2; cadherin EGF LAG seven-pas G-type receptor 2; cadherin EGF LAG seven-pass G-type receptor 2; Cadherin family member 10; cadherin, EGF LAG seven-pass G-type receptor 2; cadherin, EGF LAG seven-pass G-type receptor 2 (flamingo homolog, Drosophila); CDHF10; Celsr2; EGFL2; EGF-like protein 2; EGF-like-domain, multiple 2; epidermal growth factor-like 2; epidermal growth factor-like protein 2; flamingo; Flamingo homolog; flamingo homolog 3; Flamingo1; flamingo-like protein celsr2; FLJ34118; FLJ42737; FLJ45143; FLJ45845; KIAA0279; MEGF3; mfmi1; mKIAA0279; Multiple EGF-like domains protein 3; multiple epidermal growth factor-like domains 3; multiple epidermal growth factor-like domains protein 3; seven-pass transmembrane cadherin
Gene Symbols CELSR2
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from C-terminus of human CELSR2 / EGFL2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100061311, 100338754, 100751011, 1952, 479916, 538194, 53883, 83465
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.