Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CELSR2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981694
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1694 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1694 Supplier Bioss Supplier No. BS11818R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a member of the flamingo subfamily, part of the cadherin superfamily. The flamingo subfamily consists of nonclassic-type cadherins; a subpopulation that does not interact with catenins. The flamingo cadherins are located at the plasma membrane and have nine cadherin domains, seven epidermal growth factor-like repeats and two laminin A G-type repeats in their ectodomain. They also have seven transmembrane domains, a characteristic unique to this subfamily. It is postulated that these proteins are receptors involved in contact-mediated communication, with cadherin domains acting as homophilic binding regions and the EGF-like domains involved in cell adhesion and receptor-ligand interactions. The specific function of this particular member has not been determined.

Specifications

Antigen CELSR2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CELSR2
Gene Accession No. Q9HCU4
Gene Alias Adgrc2; adhesion G protein-coupled receptor C2; cadherin EGF LAG seven-pas G-type receptor 2; Cadherin EGF LAG seven-pass G-type receptor 2; Cadherin family member 10; cadherin, EGF LAG seven-pass G-type receptor 2; cadherin, EGF LAG seven-pass G-type receptor 2 (flamingo homolog, Drosophila); CDHF10; Celsr2; EGFL2; EGF-like protein 2; EGF-like-domain, multiple 2; epidermal growth factor-like 2; Epidermal growth factor-like protein 2; flamingo; Flamingo homolog; Flamingo homolog 3; Flamingo1; flamingo-like protein celsr2; FLJ34118; FLJ42737; FLJ45143; FLJ45845; KIAA0279; MEGF3; mfmi1; mKIAA0279; Multiple EGF-like domains protein 3; multiple epidermal growth factor-like domains 3; Multiple epidermal growth factor-like domains protein 3; seven-pass transmembrane cadherin
Gene Symbols CELSR2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CELSR2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1952
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.