Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CELT-211
GREENER_CHOICESDP

Catalog No. 795310U Shop All Novus Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-531-0U 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 79-531-0U Supplier R&D Systems™ Supplier No. 7953/10U

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Fluorescent serotonin 5HT2B receptor ligand for HTS

CELT-211 is a potent and selective fluorescent h5HT2B serotonin receptor ligand. It shows full selectivity for h5HT2B over h5HT2A, h5HT2C (Ki values is 56.3 nM for h5HT2B receptors in radioligand binding assay). Excitation and emission maxima (λ) are 589 and 616 nm, respectively. Wavelengths are compatible with use as an acceptor dye in TR-FRET assays, together with the CoraFluor™ 1 (Cat. No. 7920) TR-FRET donor.

Add 82 μL of assay buffer containing 1% of DMSO to obtain a 100 μM stock solution, and use 250 nM as a working concentration.

Specifications

Quantity 10 μg
Product Line Tocris
Target 5-HT2B
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.