Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEMIP/KIAA1199 Antibody (V98P4E1*B7), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250336FR
Description
CEMIP/KIAA1199 Monoclonal specifically detects CEMIP/KIAA1199 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
KIAA1199 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CEMIP | |
Peptide Sequence - VTLDTEDHKA peptide immunogen is identical in equine (Equus caballus), 90% identical in mouse (Mus musculus) and rat (Rattus norvegicus) | |
0.1 mL | |
Cancer | |
57214 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
V98P4E1*B7 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
CCSP1, colon cancer secreted protein 1, IR2155535, KIAA1199, TMEM2L | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction