Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEMIP Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8059R

Description
The function of this protein remains unknown.Specifications
CEMIP | |
Polyclonal | |
Unconjugated | |
Cemip | |
12H19.01.T7; 6330404C01Rik; 9930013L23Rik; AY007814; CCSP1; cell migration inducing hyaluronan binding protein; cell migration inducing hyaluronidase 1; cell migration inducing protein, hyaluronan binding; Cell migration-inducing and hyaluronan-binding protein; CEMIP; colon cancer secreted protein 1; gb:bi851057; hyaluronan-binding protein involved in hyaluronan depolymerization; HYBID; KIAA1199; LOC100353125; Protein 12H19.01.T7; protein KIAA1199; RGD1305254; TMEM2L | |
Rabbit | |
Protein A | |
RUO | |
57214 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8WUJ3 | |
Cemip | |
KLH conjugated synthetic peptide derived from human TMEM2L. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction