Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEMP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563462
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563462 Supplier Invitrogen™ Supplier No. PA563462
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSNGEKVKTI TPDVGLHQSL TSDPTVAVLR AKRAPEAHPP RSCSGSLTAR VCHMGVCQGQ GDTEDGRM Highest antigen sequence identity to the following orthologs: Mouse - 24%, Rat - 25%.

May play a role in development of the periodontium which surrounds and supports the teeth by promoting the differentiation of multi-potent cells from the periodontal ligament into cementoblasts to form the cementum (PubMed:21929512, PubMed:17509525, PubMed:21465469). Binds hydroxyapatite and may promote the biomineralization of the cementum (PubMed:19393626). Also promotes cell proliferation (PubMed:17509525, PubMed:21929512, PubMed:26011628). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEMP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CEMP1
Gene Accession No. Q6PRD7
Gene Alias Cementoblastoma-derived protein 1; cementum protein 1; Cementum protein 23; cementum protein-23; CEMP1; CP23; CP-23
Gene Symbols CEMP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CEMP1. Recombinant protein control fragment (Product #RP-98810).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 752014
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.