Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEMP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545802
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545802 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545802 Supplier Invitrogen™ Supplier No. PA545802
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RHFLNWALQQ REENSGRARR VPPVPRTAPV SKGEGSHPPQ NSNGEKVKTI Sequence homology: Human: 100%.

May play a role in development of the periodontium which surrounds and supports the teeth by promoting the differentiation of multi-potent cells from the periodontal ligament into cementoblasts to form the cementum (PubMed:21929512, PubMed:17509525, PubMed:21465469). Binds hydroxyapatite and may promote the biomineralization of the cementum (PubMed:19393626). Also promotes cell proliferation (PubMed:17509525, PubMed:21929512, PubMed:26011628). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEMP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CEMP1
Gene Accession No. Q6PRD7
Gene Alias Cementoblastoma-derived protein 1; cementum protein 1; Cementum protein 23; cementum protein-23; CEMP1; CP23; CP-23
Gene Symbols CEMP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CEMP1 (aa 136-185).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 752014
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.