Learn More
Bullard™ CEN10™ Sleek Low Profile Helmet
SDP
Catalog No. 19160563 Shop All Bullard Company Products
Click to view available options
Ventilated:
Not Vented
Vented
Color:
Dove Gray
Hi-vis Yellow
Orange
Red
White
Yellow
Bullard CEN10™ Sleek Low Profile Helmet features sleek, low-profile vented and non-vented designs, self-adjusting patented FlexGlide™ suspension system, an impact resistant helmet shell with inner foam comfort liner, and an integrated 3-point chinstrap.
  • The patented FlexGlide suspension system lets wearers auto-adjust the fit of their helmet for all day comfort
    • The CEN10 is the only auto-adjusting suspension helmet on the market, allowing even the smallest adjustments to achieve the perfect fit
  • Replaceable brow pad, comfort liner, and nape cover for ultimate utility
  • Distinctive three rib trademark
  • Manufacturer warranty: 3 years
  • Life span: 5 years
Specifications
Certifications/Compliance ANSI/ISEA Z87.1
Color Yellow
Dimensions (L x W x H) 11 x 8.75 x 6.25 in.
Liner Material EPS Foam
Material PC ABS Blend
Size Adjustable
Ventilated Not Vented
Head Style FlexGuide Head Suspension System
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.