Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CEND Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115007
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115007 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115007 Supplier Invitrogen™ Supplier No. PA5115007
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CEND1.

The protein encoded by this gene is a neuron-specific protein. The similar protein in pig enhances neuroblastoma cell differentiation in vitro and may be involved in neuronal differentiation in vivo. Multiple pseudogenes have been reported for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEND
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene CEND1
Gene Accession No. Q5FVI4, Q8N111, Q9JKC6
Gene Alias 1500001H12Rik; AI415214; BM88; BM88 antigen; C38; cell cycle exit and neuronal differentiation 1; cell cycle exit and neuronal differentiation protein 1; CEND1; RGD1309401
Gene Symbols CEND1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CEND1(Accession Q8N111), corresponding to amino acid residues P18-D68.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361675, 51286, 57754
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.