Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cenexin-1 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990908
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0908 100 μg
50-199-0909 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0908 Supplier Rockland Immunochemicals Supplier No. 200401C43
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. It is directed against, and shows specific reactivity for, human Cenexin-1 protein. A BLAST analysis was used to suggest cross-reactivity with Cenexin-1 from human, chimpanzee, mouse, rat, bovine sources based on 100% homology with the immunizing sequence. Reactivity with Cenexin-1 protein from other sources has not been determined.

Seems to be a major component of sperm tail outer dense fibers (ODF). ODFs are filamentous structures located on the outside of the axoneme in the midpiece and principal piece of the mammalian sperm tail and may help to maintain the passive elastic structures and elastic recoil of the sperm tail. May have a modulating influence on sperm motility. Functions as a general scaffold protein that is specifically localized at the distal/subdistal appendages of mother centrioles. Component of the centrosome matrix required for the localization of PLK1 and NIN to the centrosomes. Required for the formation and/or maintenance of normal CETN1 assembly.

Specifications

Antigen Cenexin-1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.53 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene ODF2
Gene Accession No. Q5BJF6
Gene Alias 84 kDa outer dense fiber protein; AI848335; cancer/testis antigen 134; Cenexin; cenexin 1; CT134; KKT4; MMTEST29; ODF2; ODF2/1; ODF2/2; Odf84; outer dense fiber of sperm tail 2; outer dense fiber of sperm tails 2; Outer dense fiber of sperm tails protein 2; outer dense fiber of sperm tails, 84-kD; Outer dense fiber protein 2; RP11-187B3.1; sperm outer dense fiber major protein 2; sperm tail structural protein; testis tissue sperm-binding protein Li 51e; testis-specific autoantigen
Gene Symbols ODF2
Host Species Rabbit
Immunogen This protein-A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a truncated recombinant protein hCenexin1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4957
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.