Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENP-A Monoclonal Antibody (5A7-2E11), QED Bioscience™
SDP

Catalog No. 502277472
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7472 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7472 Supplier Leinco Technologies Inc Supplier No. 12597100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CENP-A Monoclonal antibody specifically detects CENP-A in Human samples. It is validated for ELISA, Western Blot

Specifications

Antigen CENP-A
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5A7-2E11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene Cenpa
Gene Accession No. P49450
Gene Alias CenH3; CENP A; CENPA; CENP-A; Centromere autoantigen A; centromere protein A; centromere protein A, 17kDa; centromere-specific histone; ggCENP-A; histone H3-like centromeric protein A; RGD1563607
Host Species Mouse
Immunogen Synthetic peptide corresponding to a sequence within human CENP-A.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1058
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.