Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENP-B Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993212
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3212 100 μg
50-199-3213 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3212 Supplier Active Motif Supplier No. 61183
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CENPB is a highly conserved protein that facilitates centromere formation. It is a DNA-binding protein that is derived from transposases of the pogo DNA transposon family. It contains a helix-loop-helix DNA binding motif at the N-terminus, and a dimerization domain at the C-terminus. It is also considered a major centromere autoantigen recognized by sera from patients with anti-centromere antibodies. This gene product is a highly conserved protein that facilitates centromere formation. It is a DNA-binding protein that is derived from transposases of the pogo DNA transposon family. It contains a helix-loop-helix DNA binding motif at the N-terminus, and a dimerization domain at the C-terminus. The DNA binding domain recognizes and binds a 17-bp sequence (CENP-B box) in the centromeric alpha satellite DNA. This protein is proposed to play an important role in the assembly of specific centromere structures in interphase nuclei and on mitotic chromosomes and is considered a major centromere autoantigen recognized by sera from patients with anti-centromere antibodies.

Specifications

Antigen CENP-B
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene CENPB
Gene Accession No. P07199
Gene Alias Cenpb; Cenp-b; CENP-B protein; centromere autoantigen B; Centromere protein B; centromere protein B, 80kDa; Major centromere autoantigen B
Gene Symbols CENPB
Host Species Rabbit
Immunogen This CENP-B antibody was raised against a recombinant protein corresponding to the C-terminal region of human CENP-B.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1059
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.