Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENP-E Monoclonal Antibody (1H12), Active Motif™
SDP

Mouse Monoclonal Antibody

$132.55 - $397.65

Specifications

Antigen CENP-E
Clone 1H12
Concentration 1 mg/mL
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-2740
SDP
Encompass
View Documents
Active Motif
39620
10 μg
Each for $132.55
Only null left
Add to Cart
 
50-199-2739
SDP
Encompass
View Documents
Active Motif
39619
200 μg
Each for $397.65
Only null left
Add to Cart
 
Description

Description

14-3-3 sigma (stratifin, SFN, epithelial cell marker protein 1) is a member of a highly conserved family of 14-3-3 proteins that are present in all eukaryotic organisms. There are 7 known mammalian 14-3-3 isoforms epsilon, beta, zeta, gamma, theta, tau and they play important roles in many biological activities by binding to and altering the subcellular localization and/or stability of key molecules in various signaling cascades. 14-3-3 sigma was originally characterized as a human mammary epithelium marker and later rediscovered as an important molecule for cell cycle checkpoint regulation. Recently it has been reported that 14-3-3 sigma may serve as a prognosis marker predicting survival of pancreatic cancer patient treatment. 14-3-3 sigmais the only 14-3-3 isoform induced by the tumor suppressor protein p53, in response to γ-irradiation and other DNA-damaging agents. 14-3-3 sigmais a p53-regulated inhibitor of G2/M progression and acts as a tumor suppressor gene that is inactivated by methylation of its 5' CpG islands in epithelial tumor cells. Two isoforms of human 14-3-3 sigma are produced by alternative splicing.
Specifications

Specifications

CENP-E
1 mg/mL
Monoclonal
Liquid
RUO
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
CENPE; CENP-E; Centromere autoantigen E (312kD); Centromere protein E; centromere protein E, 312kDa; Centromere-associated protein E; KIF10; kinesin family member 10; Kinesin-7; Kinesin-related protein CENPE; MCPH13; PPP1R61; protein phosphatase 1, regulatory subunit 61
CENPE
IgG
Protein G
Antibody
1H12
Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Human, Mouse
Q02224, Q6RT24
1062, 229841
This CENP-E antibody was raised against full-length human CENP-E protein.
Primary
-20°C, Avoid Freeze/Thaw Cycles
CENPE
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.